Learn More
CTTN Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CTTN.
Brand: Abnova PAB25120.100uL
Description
This gene is overexpressed in breast cancer and squamous cell carcinomas of the head and neck. The encoded protein is localized in the cytoplasm and in areas of the cell-substratum contacts. This gene has two roles: (1) regulating the interactions between components of adherens-type junctions and (2) organizing the cytoskeleton and cell adhesion structures of epithelia and carcinoma cells. During apoptosis, the encoded protein is degraded in a caspase-dependent manner. The aberrant regulation of this gene contributes to tumor cell invasion and metastasis. Two splice variants that encode different isoforms have been identified for this gene. [provided by RefSeq
Specifications
| CTTN | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| CTTN | |
| CTTN | |
| A synthetic peptide corresponding to CTTN. | |
| 100 μL | |
| Primary | |
| CTTN polyclonal antibody detects endogenous levels of CTTN protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CTTN. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| EMS1/FLJ34459 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 2017 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only