Learn More
CTSS Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CTSS.
Brand: Abnova PAB28624.100uL
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The encoded protein can function as an elastase over a broad pH range in alveolar macrophages. Transcript variants utilizing alternative polyadenylation signals exist for this gene. [provided by RefSeq
Sequence: NGGFMTTAFQYIIDNKGIDSDASYPYKAMDQKCQYDSKYRAATCSKYTELPYGREDVLKEAVANKGPVSVGVDARHPSFFLYRSGVYYEPSCTQNVNHGVLVVGYGDLNGKEYWLVKNSWGHNFGEEGYIRMARNKGNHCGSpecifications
CTSS | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant CTSS. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
P25774 | |
CTSS | |
Recombinant protein corresponding to amino acids of human CTSS. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
CTSS | |
MGC3886 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
1520 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
For Research Use Only