missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTNNA1/CTNNA2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CTNNA1/CTNNA2.
Brand: Abnova PAB27026.100uL
This item is not returnable.
View return policy
Specifications
| CTNNA1/CTNNA2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| P35221 (Gene ID : 1495);P26232 (Gene ID : 1496) | |
| CTNNA1 | |
| A synthetic peptide corresponding to human CTNNA1/CTNNA2. | |
| 100 μL | |
| Primary | |
| CTNNA1/CTNNA2 polyclonal antibody detects endogenous levels of CTNNA1/CTNNA2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CTNNA1/CTNNA2. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| CAP102/FLJ36832 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1495/1496 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction