Learn More
CRYL1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CRYL1.
Brand: Abnova PAB23654.100uL
Description
The uronate cycle functions as an alternative glucose metabolic pathway, accounting for about 5% of daily glucose catabolism. The product of this gene catalyzes the dehydrogenation of L-gulonate into dehydro-L-gulonate in the uronate cycle. The enzyme requires NAD(H) as a coenzyme, and is inhibited by inorganic phosphate. A similar gene in the rabbit is thought to serve a structural role in the lens of the eye. [provided by RefSeq
Sequence: IRNALENIRKEMKLLEQAGSLKGSLSVEEQLSLISGCPNIQEAVEGAMHIQECVPEDLELKKKIFAQLDSIIDDRVILSSSTSCLMSpecifications
| CRYL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CRYL1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9Y2S2 | |
| CRYL1 | |
| Recombinant protein corresponding to amino acids of human CRYL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
| CRYL1 | |
| GDH/MGC149525/MGC149526/lambda-CRY | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51084 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only