missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CRP.
Brand: Abnova PAB7968.100ug
This item is not returnable.
View return policy
Specifications
| CRP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CRP. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| MGC149895/MGC88244/PTX1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 1401 | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CRP | |
| CRP | |
| A synthetic peptide corresponding to amino acids at C-terminus of human CRP. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur