Learn More
CRLF2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CRLF2.
Brand: Abnova PAB13267.100ug
Description
Cytokine signals are mediated through specific receptor complexes, the components of which are mostly members of the type I cytokine receptor family. Type I cytokine receptors share conserved structural features in their extracellular domain. Receptor complexes are typically heterodimeric, consisting of alpha chains, which provide ligand specificity, and beta (or gamma) chains, which are required for the formation of high-affinity binding sites and signal transduction.[supplied by OMIM
Specifications
| CRLF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CRLF2. | |
| In PBS (0.02% sodium azide) | |
| Q9HC73 | |
| CRLF2 | |
| A synthetic peptide corresponding to internal region 19 amino acids of human CRLF2. | |
| RUO | |
| 64109 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CRLF2 | |
| CRL2/CRLF2Y/TSLPR | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only