Learn More
CREM, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CREM
Brand: Abnova PAB28489.100uL
Description
This gene encodes a bZIP transcription factor that binds to the cAMP responsive element found in many viral and cellular promoters. It is an important component of cAMP-mediated signal transduction during the spermatogenetic cycle, as well as other complex processes. Alternative promoter and translation initiation site usage allows this gene to exert spatial and temporal specificity to cAMP responsiveness. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene, with some of them functioning as activators and some as repressors of transcription. [provided by RefSeq
Sequence: QHDGSITASLTESKSAHVQTQTGQNSIPALAQVAAIAETDESAESEGVIDSHKRREILSRRPSYRKILNELSSDVPGVPKIEEERSEEEGTPPSIATMAVPTSIYQTSTGQYIAIAQGGTIQISNPGSDSpecifications
| CREM | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CREM | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| E9PBM5 | |
| CREM | |
| Recombinant protein corresponding to amino acids of human CREM | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:500-1:1000) Western Blot(1:250-1:500) Immunofluorescence(1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| CREM | |
| ICER/MGC111110/MGC17881/MGC41893/hCREM-2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1390 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only