Learn More
CPE, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CPE.
Brand: Abnova PAB17650.100ug
Description
This gene encodes a carboxypeptidase that cleaves C-terminal amino acid residues and is involved in the biosynthesis of peptide hormones and neurotransmitters, including insulin. It is a peripheral membrane protein. The protein specifically binds regulated secretory pathway proteins, including prohormones, but not constitutively secreted proteins. Mutations in this gene are implicated in type II diabetes. [provided by RefSeq
Specifications
| CPE | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CPE. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| P16870 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total CPE protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| CPE | |
| CPE | |
| A synthetic peptide corresponding to internal of human CPE. | |
| RUO | |
| 1363 | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only