Learn More
CORO7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CORO7.
Brand: Abnova PAB25716.100ug
Description
Coronins, such as CORO7, constitute an evolutionarily conserved family of WD-repeat actin-binding proteins. CORO7 plays a role in Golgi complex morphology and function (Rybakin et al., 2004, 2006 [PubMed 15327992] [PubMed 16905771]).[supplied by OMIM
Specifications
| CORO7 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CORO7 | |
| 0610011B16Rik/CRN7/FLJ22021/FLJ44188/POD1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 79585 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CORO7. | |
| In PBS (0.02% sodium azide) | |
| NP_078811 | |
| CORO7 | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human CORO7. | |
| 100 μg | |
| Primary | |
| Multiple isoforms of CORO7 are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only