Learn More
COQ9, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant COQ9.
Brand: Abnova PAB23716.100uL
Description
Coenzyme Q10 (CoQ10), or ubiquinone, is a mobile lipophilic electron carrier critical for electron transfer by the mitochondrial inner membrane respiratory chain. COQ9 is 1 of several enzymes involved in biosynthesis of CoQ10 and likely functions in modification of the benzoquinone ring (Duncan et al., 2009 [PubMed 19375058]).[supplied by OMIM
Sequence: DQSTDFNWYTRRAMLAAIYNTTELVMMQDSSPDFEDTWRFLENRVNDAMNMGHTAKQVKSTGEALVQGLMGAAVTLKNLSpecifications
| COQ9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant COQ9. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| O75208 | |
| COQ9 | |
| Recombinant protein corresponding to amino acids of human COQ9. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| COQ9 | |
| C16orf49/DKFZp434K046 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57017 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only