Learn More
COPS5 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of COPS5.
Brand: Abnova PAB26896.100uL
Description
The protein encoded by this gene is one of the eight subunits of COP9 signalosome, a highly conserved protein complex that functions as an important regulator in multiple signaling pathways. The structure and function of COP9 signalosome is similar to that of the 19S regulatory particle of 26S proteasome. COP9 signalosome has been shown to interact with SCF-type E3 ubiquitin ligases and act as a positive regulator of E3 ubiquitin ligases. This protein is reported to be involved in the degradation of cyclin-dependent kinase inhibitor CDKN1B/p27Kip1. It is also known to be an coactivator that increases the specificity of JUN/AP1 transcription factors. [provided by RefSeq
Specifications
| COPS5 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| COPS5 | |
| CSN5/JAB1/MGC3149/MOV-34/SGN5 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 10987 | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of COPS5. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| Q92905 | |
| COPS5 | |
| A synthetic peptide corresponding to human COPS5. | |
| 100 μL | |
| Primary | |
| COPS5 polyclonal antibody detects endogenous levels of COPS5 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only