missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I and III Rabbit anti-Bovine, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Collagen I and III.
Brand: Abnova PAB13716.1mL
This item is not returnable.
View return policy
Specifications
| Collagen I and III | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native Collagen I and III. | |
| Lyophilized from purified antibody | |
| COL1A1 | |
| Native purified bovine Collagen I and III. | |
| RUO | |
| 282187/282188/510833 | |
| Bovine | |
| Lyophilized |
| Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Immunohistochemistry (Frozen sections) (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| MGC128258 | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Recognizes calf collagen type I and III. | |
| Store at 4°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction