Learn More
COLEC11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant COLEC11.
Brand: Abnova PAB22718.100uL
Description
COLEC11 is a member of the collectin family of C-type lectins, which contain a collagen-like domain and a carbohydrate recognition domain, and play a role in host-defense (Keshi et al., 2006 [PubMed 17179669]).[supplied by OMIM
Sequence: INDLEKEGAFVYSDHSPMRTFNKWRSGEPNNAYDEEDCVEMVASGGWNDVACHTTMYFMCEFDKENMSpecifications
| COLEC11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant COLEC11. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9BWP8 | |
| COLEC11 | |
| Recombinant protein corresponding to amino acids of human COLEC11. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| COLEC11 | |
| CL-K1-I/CL-K1-II/CL-K1-IIa/CL-K1-IIb/CLK1/DKFZp686N1868/MGC3279 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 78989 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only