missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL5A1/COL5A2/COL5A3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native COL5A1/COL5A2/COL5A3.
Brand: Abnova PAB13507.100uL
This item is not returnable.
View return policy
Specifications
| COL5A1/COL5A2/COL5A3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native COL5A1/COL5A2/COL5A3. | |
| Lyophilized from PBS | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody reacts with human collagen type V 100% binding in RIA at 1:100 dilution, cross-reactivity with human collagen type I, III and IV <0.1%; human fibronectin and laminin <0.1%. | |
| Store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA) | |
| Unconjugated | |
| ELISA (>1:200) Immunohistochemistry (1:500) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user. | |
| COL5A1 | |
| Native purified COL5A1/COL5A2/COL5A3 from human placenta. | |
| RUO | |
| 1289/1290/50509 | |
| Bovine, Human, Pig | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction