missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL4A3 Rabbit anti-Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native COL4A3.
Brand: Abnova PAB13506.100uL
This item is not returnable.
View return policy
Specifications
| COL4A3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native COL4A3. | |
| Lyophilized from phosphate buffer | |
| COL4A3 | |
| Native purified COL4A3 from pig kidney tissue. | |
| RUO | |
| 397502 | |
| Pig | |
| Lyophilized |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA) | |
| Unconjugated | |
| ELISA (>1:200) Immunohistochemistry (1:500) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user. | |
| COL4A3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody reacts with porcine collagen type IV 100%, cross-reactivity with porcine collagens type I, II, and III <0.1%; porcine elastin, fibronectin and laminin <0.1. | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction