Learn More
COL2A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of COL2A1.
Brand: Abnova PAB3065.400uL
Description
This gene encodes the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia. There are two transcripts identified for this gene. [provided by RefSeq
Specifications
| COL2A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of COL2A1. | |
| In PBS (0.09% sodium azide) | |
| NP_001835;P02458 | |
| COL2A1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human COL2A1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| COL2A1 | |
| ANFH/AOM/COL11A3/MGC131516/SEDC | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 1280 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only