Learn More
COL17A1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant COL17A1.
Brand: Abnova PAB28148.100uL
Description
This gene encodes the alpha chain of type XVII collagen. Unlike most collagens, collagen XVII is a transmembrane protein. Collagen XVII is a structural component of hemidesmosomes, multiprotein complexes at the dermal-epidermal basement membrane zone that mediate adhesion of keratinocytes to the underlying membrane. Mutations in this gene are associated with both generalized atrophic benign and junctional epidermolysis bullosa. Two homotrimeric forms of type XVII collagen exist. The full length form is the transmembrane protein. A soluble form, referred to as either ectodomain or LAD-1, is generated by proteolytic processing of the full length form. [provided by RefSeq
Sequence: DGTEVTERIVTETVTTRLTSLPPKGGTSNGYAKTASLGGGSRLEKQSLTHGSSGYINSTGSTRGHASTSSYRRAHSPASTLPNSPGSTFERKTHVTRSpecifications
| COL17A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant COL17A1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| Q9UMD9 | |
| COL17A1 | |
| Recombinant protein corresponding to amino acids of recombinant COL17A1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| COL17A1 | |
| BA16H23.2/BP180/BPAG2/FLJ60881/KIAA0204/LAD-1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1308 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only