Learn More
COL12A1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of COL12A1.
Brand: Abnova PAB17518.100ug
Description
This gene encodes the alpha chain of type XII collagen, a member of the FACIT (fibril-associated collagens with interrupted triple helices) collagen family. Type XII collagen is a homotrimer found in association with type I collagen, an association that is thought to modify the interactions between collagen I fibrils and the surrounding matrix. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
| COL12A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of COL12A1. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q99715 | |
| COL12A1 | |
| A synthetic peptide corresponding to internal of human COL12A1. | |
| RUO | |
| 1303 | |
| Human | |
| Liquid |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| COL12A1 | |
| BA209D8.1/COL12A1L/DJ234P15.1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total COL12A1 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only