Learn More
CNRIP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CNRIP1.
Brand: Abnova PAB27423.100ug
Description
This gene encodes a G-protein coupled receptor which interacts with the C-terminal tail of cannabinoid receptor 1. This receptor plays a role in synaptic plasticity, analgesia, appetite, and neuroprotection. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Specifications
| CNRIP1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| CNRIP1 | |
| C2orf32/CRIP1/DKFZp566K1924 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 25927 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CNRIP1. | |
| In PBS (0.02% sodium azide) | |
| NP_056278 | |
| CNRIP1 | |
| A synthetic peptide corresponding to 15 amino acids at internal region of human CNRIP1. | |
| 100 μg | |
| Primary | |
| At least two isoforms of CNRIP1 are known to exist; this antibody will detect both isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only