Learn More
CNR2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyconal antibody raised against synthetic peptide of CNR2.
Brand: Abnova PAB28336.50ug
Description
The cannabinoid delta-9-tetrahydrocannabinol is the principal psychoactive ingredient of marijuana. The proteins encoded by this gene and the cannabinoid receptor 1 (brain) (CNR1) gene have the characteristics of a guanine nucleotide-binding protein (G-protein)-coupled receptor for cannabinoids. They inhibit adenylate cyclase activity in a dose-dependent, stereoselective, and pertussis toxin-sensitive manner. These proteins have been found to be involved in the cannabinoid-induced CNS effects (including alterations in mood and cognition) experienced by users of marijuana. The cannabinoid receptors are members of family 1 of the G-protein-coupled receptors. [provided by RefSeq
Specifications
| CNR2 | |
| Polyclonal | |
| Rabbit polyconal antibody raised against synthetic peptide of CNR2. | |
| In PBS (0.09% sodium azide) | |
| P34972 | |
| CNR2 | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human CNR2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (11 ug/ml) The optimal working dilution should be determined by the end user. | |
| CNR2 | |
| CB2/CX5 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 1269 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only