Learn More
CMTM6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CMTM6.
Brand: Abnova PAB21992.100uL
Description
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq
Sequence: KRQESQLRKPENTTRAEALTEPLNASpecifications
| CMTM6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CMTM6. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NX76 | |
| CMTM6 | |
| Recombinant protein corresponding to amino acids of human CMTM6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| CMTM6 | |
| CKLFSF6/FLJ20396/PRO2219 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54918 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only