missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMKLR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CMKLR1.
Brand: Abnova PAB26144.50ug
Additional Details : Weight : 0.02000kg
Specifications
CMKLR1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CMKLR1. | |
In PBS (0.09% sodium azide) | |
Q99788 | |
CMKLR1 | |
A synthetic peptide corresponding to 19 amino acids at C-terminus of human CMKLR1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
CMKLR1 | |
CHEMERINR/ChemR23/DEZ/MGC126105/MGC126106 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
1240 | |
Human | |
Liquid |
For Research Use Only