missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTA/CLTB, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CLTA/CLTB.
Brand: Abnova PAB14385.200uL
This item is not returnable.
View return policy
Description
Sequence: EEDPAAAFLAQQESEIAGIENDSpecifications
| CLTA/CLTB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CLTA/CLTB. | |
| Lyophilized from serum | |
| LCA | |
| Rabbit | |
| 200 μL | |
| Primary | |
| This antibody recognizes both CLCa and CLCb isoforms of human CLC. This antibody recognizes bands migrating around 30 KDa. | |
| Store at 4°C or -20°C on dry atmosphere. After reconstitution with water and glycerol (1:1), store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) Immunofluorescence (1:100-1:500) The optimal working dilution should be determined by the end user. | |
| P09496 | |
| CLTA | |
| A synthetic peptide corresponding to human CLTA/CLTB. | |
| RUO | |
| 1211/1212 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction