Learn More
CLDN10 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CLDN10.
Brand: Abnova PAB27032.100uL
Description
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| CLDN10 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| CLDN10 | |
| CPETRL3/OSP-L | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9071 | |
| Human, Mouse | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CLDN10. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| P78369 | |
| CLDN10 | |
| A synthetic peptide corresponding to human CLDN10. | |
| 100 μL | |
| Primary | |
| CLDN10 polyclonal antibody detects endogenous levels of CLDN10 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only