Learn More
CLCN4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CLCN4.
Brand: Abnova PAB22540.100uL
Description
The CLCN family of voltage-dependent chloride channel genes comprises nine members (CLCN1-7, Ka and Kb) which demonstrate quite diverse functional characteristics while sharing significant sequence homology. Chloride channel 4 has an evolutionary conserved CpG island and is conserved in both mouse and hamster. This gene is mapped in close proximity to APXL (Apical protein Xenopus laevis-like) and OA1 (Ocular albinism type I), which are both located on the human X chromosome at band p22.3. The physiological role of chloride channel 4 remains unknown but may contribute to the pathogenesis of neuronal disorders. [provided by RefSeq
Sequence: VVSRDSERLIGFAQRRELILAIKNARQRQEGIVSNSIMYFTEEPPELPANSPHPLKLRRILNLSSpecifications
| CLCN4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CLCN4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P51793 | |
| CLCN4 | |
| Recombinant protein corresponding to amino acids of human CLCN4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| CLCN4 | |
| CLC4/ClC-4/ClC-4A/MGC163150 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1183 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only