Learn More
CLC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CLC.
Brand: Abnova PAB23850.100uL
Description
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene is a lysophospholipase expressed in eosinophils and basophils. It hydrolyzes lysophosphatidylcholine to glycerophosphocholine and a free fatty acid. This protein may possess carbohydrate or IgE-binding activities. It is both structurally and functionally related to the galectin family of beta-galactoside binding proteins. It may be associated with inflammation and some myeloid leukemias. [provided by RefSeq
Sequence: LACFLNEPYLQVDFHTEMKEESDIVFHFQVCFGRRVVMNSREYGAWKQQVESKNMPFQDGQEFELSISVLPDKYQVMSpecifications
| CLC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CLC. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q05315 | |
| CLC | |
| Recombinant protein corresponding to amino acids of human CLC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:2500-1:5000) The optimal working dilution should be determined by the end user. | |
| CLC | |
| LGALS10/LPPL_HUMAN/MGC149659 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1178 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only