Learn More
CHUK Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CHUK.
Brand: Abnova PAB28431.100uL
Description
This gene encodes a member of the serine/threonine protein kinase family. The encoded protein, a component of a cytokine-activated protein complex that is an inhibitor of the essential transcription factor NF-kappa-B complex, phosphorylates sites that trigger the degradation of the inhibitor via the ubiquination pathway, thereby activating the transcription factor. [provided by RefSeq
Sequence: HTVQSQDRVLKELFGHLSKLLGCKQKIIDLLPKVEVALSNIKEADNTVMFMQGKRQKEIWHLLKIACTQSSARSLVGSSLEGAVTPQTSAWLPPTSAEHDHSLSCVVTPQDGETSAQMIEENLNCLGHLSTIIHEANEEQGNSMMNSpecifications
| CHUK | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CHUK. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| O15111 | |
| CHUK | |
| Recombinant protein corresponding to amino acids of recombinant CHUK. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:200-1:500) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| CHUK | |
| IKBKA/IKK-alpha/IKK1/IKKA/NFKBIKA/TCF16 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1147 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only