Learn More
CHRM2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CHRM2.
Brand: Abnova PAB26085.50ug
Description
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine to these receptors and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 2 is involved in mediation of bradycardia and a decrease in cardiac contractility. Multiple alternatively spliced transcript variants have been described for this gene. [provided by RefSeq
Specifications
| CHRM2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CHRM2. | |
| In PBS (0.09% sodium azide) | |
| P08172 | |
| CHRM2 | |
| A synthetic peptide corresponding to cytoplasmic domain of human CHRM2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-15 ug/mL) The optimal working dilution should be determined by the end user. | |
| CHRM2 | |
| FLJ43243/HM2/MGC120006/MGC120007 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 1129 | |
| Bovine, Canine, Equine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only