Learn More
CHN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CHN2.
Brand: Abnova PAB21183.100uL
Description
This gene is a member of the chimerin family and encodes a protein with a phorbol-ester/DAG-type zinc finger, a Rho-GAP domain and an SH2 domain. This protein has GTPase-activating protein activity that is regulated by phospholipid binding and binding of diacylglycerol (DAG) induces translocation of the protein from the cytosol to the Golgi apparatus membrane. The protein plays a role in the proliferation and migration of smooth muscle cells. Decreased expression of this gene is associated with high-grade gliomas and breast tumors, and increased expression of this gene is associated with lymphomas. Mutations in this gene have been associated with schizophrenia in men. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: ASSNSSLSGSSVSSDAEEYQPPIWKSYLYQLQQEAPRPKRIICPREVENRPKYSpecifications
| CHN2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CHN2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P52757 | |
| CHN2 | |
| Recombinant protein corresponding to amino acids of human CHN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| CHN2 | |
| ARHGAP3/BCH/MGC138360/RHOGAP3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1124 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only