Learn More
CHN1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CHN1.
Brand: Abnova PAB8940.100ug
Description
This gene encodes GTPase-activating protein for p21-rac and a phorbol ester receptor. It plays an important role in ocular motor axon pathfinding. Heterozygous missense mutations in this gene cause Duane's retraction syndrome 2 (DURS2). Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: REPAYKKHMPVLKETHDERDSpecifications
| CHN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CHN1. | |
| In buffer containing 0.02% sodium azide | |
| ARHGAP2/CHN/DURS2/RHOGAP2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| CHN1 | |
| CHN1 | |
| A synthetic peptide corresponding to amino acids 153-172 of human CHN1. | |
| RUO | |
| 1123 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only