Learn More
CHM, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CHM.
Brand: Abnova PAB20184.100uL
Description
This gene encodes component A of the RAB geranylgeranyl transferase holoenzyme. In the dimeric holoenzyme, this subunit binds unprenylated Rab GTPases and then presents them to the catalytic Rab GGTase subunit for the geranylgeranyl transfer reaction. Rab GTPases need to be geranylgeranyled on either one or two cysteine residues in their C-terminus to localize to the correct intracellular membrane. Mutations in this gene are a cause of choroideremia; also known as tapetochoroidal dystrophy (TCD). This X-linked disease is characterized by progressive dystrophy of the choroid, retinal pigment epithelium and retina. Alternative splicing results in multiple transcript variants encoding different isoforms
Sequence: SRKDKTIQHVEVFCYASQDLHEDVEEAGALQKNHALVTSANSTEAADSAFLPTEDESLSTMSCEMLTEQTPSSDPENALEVNGAEVTGEKENHCDDKTCVPSTSAEDMSENVPIAEDTTEQPKKNRITYSQIIKEGSpecifications
| CHM | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CHM. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P24386 | |
| CHM | |
| Recombinant protein corresponding to amino acids of human CHM. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| CHM | |
| DXS540/FLJ38564/GGTA/HSD-32/MGC102710/REP-1/TCD | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1121 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only