missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Chicken IgY (H+L) Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB7299
This item is not returnable.
View return policy
Description
Buffer: Phosphate Buffered Saline (PBS) containing 0.2%BSA.IgY (H+L) Polyclonal specifically detects IgY (H+L) in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IgY (H+L) | |
| Polyclonal | |
| HRP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| By immunoelectrophoresis and ELISA this reacts specifically with rabbit IgG and with light chains common to other rabbit immunoglobulins. No was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species. | |
| Store at 4C. Do not freeze. | |
| Chicken |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:10000, ELISA, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry/Immunofluorescence 1:200 - 1:5000, Immunohistochemistry-Paraffin | |
| Chicken IgG (IgY)-heavy and light chain | |
| 1 mL | |
| Secondary | |
| Chicken | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction