Learn More
Invitrogen™ Rabbit anti-Chicken IgY (H+L) Secondary Antibody, FITC
Rabbit Polyclonal Secondary Antibody
Marca:  Invitrogen™ 31501
Código de producto. 11809160
  • £162.00 / 1500µg
Este artículo no se puede devolver. Vea la política de devoluciones
Descripción
Descripción
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Antibody Specificity: This antibody reacts with the heavy chains on chicken IgY and with the light chains common to most chicken immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins using immunoelectrophoresis. However, these antibodies may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 2-8°C until opened. Restore to 1.1 mL with distilled water (1.5 mg/mL after restoration). Centrifuge product if it is not clear upon standing for 1-2 hours at room temperature. To judge clarity, draw the product into a pasteur pipette. Product may be stored for several weeks at 2-8°C as undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorophore: Fluorescein-5-isothiocyanate (FITC-isomer 1) Amax = 492 nm; Emax = 520 nm Fluorophore/Protein: 8.5 μg/mg; 3.2 moles FITC per mole IgG Country of Origin: USA.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Chicken IgY (H+L)
Polyclonal
FITC
Rabbit
1.5 mg
Secondary
4°C, store in dark
Lyophilized
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
1.5 mg/mL
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Antigen affinity chromatography
RUO
Chicken
Secondary Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado