Learn More
Rabbit anti-Chicken IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB10506.1500ug
Product Code. 16202850
  • £349.00 / 1500µg
Specifications
Rabbit anti-Chicken IgG whole molecule
Polyclonal
Rabbit anti-chicken IgG recognizes chicken IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicin sulfate)
Chicken IgG whole molecule
1500 μg
Chicken
Lyophilized
Dot Blot, ELISA, Immunohistochemistry, Western Blot
HRP
The optimal working dilution should be determined by the end user.
Rabbit
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.