missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD7 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CHD7.
Brand: Abnova PAB25712.100ug
This item is not returnable.
View return policy
Description
This gene encodes a protein that contains several helicase family domains. Mutations in this gene have been found in some patients with the CHARGE syndrome. [provided by RefSeq
Specifications
| CHD7 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CHD7 | |
| FLJ20357/FLJ20361/IS3/KAL5/KIAA1416 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 55636 | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CHD7. | |
| In PBS (0.02% sodium azide) | |
| Q9P2D1 | |
| CHD7 | |
| A synthetic peptide corresponding to 18 amino acids at N-terminus of human CHD7. | |
| 100 μg | |
| Primary | |
| Multiple isoforms of CHD7 are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction