missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAT Rabbit anti-Guinea pig, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CHAT.
Brand: Abnova PAB14313.100uL
This item is not returnable.
View return policy
Description
Sequence: GLFSSYRLPGHTQDTLVAQKSSSpecifications
| CHAT | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CHAT. | |
| Lyophilized from PBS | |
| ChoAcTase | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antiserum stains cholinergic neurons in guinea-pig and rabbit. | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:3000) Western Blot (1:1000-1:3000) ELISA (1:1000-1:3000) The optimal working dilution should be determined by the end user. | |
| CHAT | |
| CHAT | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 167-188 of pig CHAT. | |
| RUO | |
| 396896 | |
| Guinea Pig, Mouse, Pig, Rabbit, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction