missing translation for 'onlineSavingsMsg'
Learn More

CFL1/CFL2 (phospho Y88), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of CFL1/CFL2.

Brand:  Abnova PAB25901.100ug

Product Code. 16086077

  • £349.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: A-T-Yp-E-T
Specifications

Specifications

CFL1/CFL2
Polyclonal
Unconjugated
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
P23528 (Gene ID : 1072);Q9Y281 (Gene ID : 1073)
CFL1
Synthetic phosphopeptide corresponding to residues surrounding Y88 of human CFL1/CFL2.
100 μg
Primary
Human, Mouse, Rat
Liquid
Immunohistochemistry (PFA fixed)
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CFL1/CFL2.
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
CFL
Rabbit
Affinity chromatography
RUO
1072/1073
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only