Learn More
CEP350 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CEP350.
Brand: Abnova PAB17661.100ug
Description
The product of this gene is a large protein with a CAP-Gly domain typically found in cytoskeleton-associated proteins. The encoded protein primarily localizes to the centrosome, a nonmembraneous organelle that functions as the major microtubule-organizing center in animal cells. The encoded protein directly interacts with another large centrosomal protein and is required to anchor microtubules at the centrosome. It is also implicated in the regulation of a class of nuclear hormone receptors in the nucleus. Several alternatively spliced transcript variants have been found, but their full-length nature has not been determined. [provided by RefSeq
Specifications
| CEP350 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CEP350. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q5VT06 | |
| CEP350 | |
| A synthetic peptide corresponding to internal of human CEP350. | |
| RUO | |
| 9857 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| CEP350 | |
| CAP350/FLJ38282/FLJ44058/GM133/KIAA0480 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total CEP350 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only