Learn More
CEACAM3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CEACAM3.
Brand: Abnova PAB20527.100uL
Description
This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq
Sequence: NVTQNDIGFYTLQVIKSDLVNEEATGQFHVYQESpecifications
| CEACAM3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CEACAM3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P40198 | |
| CEACAM3 | |
| Recombinant protein corresponding to amino acids of human CEACAM3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| CEACAM3 | |
| CD66D/CEA/CGM1/MGC119875/W264/W282 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1084 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only