Learn More
CDH7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDH7.
Brand: Abnova PAB3132.400uL
Description
This gene is a type II classical cadherin from the cadherin superfamily. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Cadherins mediate cell-cell binding in a homophilic manner, contributing to the sorting of heterogeneous cell types and the maintenance of orderly structures. Alternative splicing in the 5' UTR of this gene yields variant transcripts encoding the same protein. [provided by RefSeq]
Specifications
| CDH7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDH7. | |
| In PBS (0.09% sodium azide) | |
| NP_387450;Q9ULB5 | |
| CDH7 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CDH7. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user. | |
| CDH7 | |
| CDH7L1 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 1005 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only