Learn More
CDH26 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDH26.
Brand: Abnova PAB17494.100ug
Description
Cadherins are a family of adhesion molecules that mediate Ca2+-dependent cell-cell adhesion in all solid tissues and modulate a wide variety of processes, including cell polarization and migration. Cadherin domains occur as repeats in the extracellular region and are thought to contribute to the sorting of heterogeneous cell types and the maintenance of orderly structures such as epithelium. This gene encodes a cadherin domain-containing protein whose specific function has not yet been determined. Alternative splicing occurs at this locus and two transcript variants, encoding distinct proteins, have been identified. [provided by RefSeq]
Specifications
| CDH26 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDH26. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q8IXH8 | |
| CDH26 | |
| A synthetic peptide corresponding to internal of human CDH26. | |
| RUO | |
| 60437 | |
| Human | |
| Liquid |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| CDH26 | |
| VR20 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total CDH26 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only