Learn More
CDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDH1.
Brand: Abnova PAB7893.100uL
Description
This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. [provided by RefSeq
Specifications
| CDH1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDH1. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| P12830 | |
| CDH1 | |
| A synthetic peptide corresponding to C-terminus of human CDH1. | |
| RUO | |
| 999 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| CDH1 | |
| Arc-1/CD324/CDHE/ECAD/LCAM/UVO | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This sequence is conserved in rat and mouse E-cadherin, and has low homology to other cadherin family members. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only