Learn More
CDC25B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDC25B.
Brand: Abnova PAB2829.400uL
Description
CDC25B is a member of the CDC25 family of phosphatases. CDC25B activates the cyclin dependent kinase CDC2 by removing two phosphate groups and it is required for entry into mitosis. CDC25B shuttles between the nucleus and the cytoplasm due to nuclear localization and nuclear export signals. The protein is nuclear in the M and G1 phases of the cell cycle and moves to the cytoplasm during S and G2. CDC25B has oncogenic properties, although its role in tumor formation has not been determined. Multiple transcript variants for this gene exist. [provided by RefSeq
Spécification
| CDC25B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDC25B. | |
| In PBS (0.09% sodium azide) | |
| NP_068660;P30305 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 994 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| CDC25B | |
| CDC25B | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human CDC25B. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only