Learn More
CDC16 (phospho T580), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDC16.
Brand: Abnova PAB10325.100ug
Description
This gene encodes a component protein of the APC complex, which is composed of eight proteins and functions as a protein ubiquitin ligase. The APC complex is a cyclin degradation system that governs exit from mitosis. Each component protein of the APC complex is highly conserved among eukaryotic organisms. This protein and two other APC complex proteins, CDC23 and CDC27, contain a tetratricopeptide repeat (TPR), a protein domain that may be involved in protein-protein interaction. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Specifications
| CDC16 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDC16. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Q13042 | |
| CDC16 | |
| Synthetic phosphopeptide corresponding to residues surrounding T580 of human CDC16. | |
| RUO | |
| 8881 | |
| Human | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| ELISA (1:1000-1:10000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| CDC16 | |
| APC6 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody is specific to phosphorylated human APC6 protein at the pT580 residue. This antibody is expected to react with all isoforms of this protein. Minimal reactivity is expected with the non-phosphorylated form of the protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only