Learn More
CD40LG Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CD40LG.
Brand: Abnova PAB28325.100uL
Description
The protein encoded by this gene is expressed on the surface of T cells. It regulates B cell function by engaging CD40 on the B cell surface. A defect in this gene results in an inability to undergo immunoglobulin class switch and is associated with hyper-IgM syndrome. [provided by RefSeq
Sequence: LTVKRQGLYYIYAQVTFCSNREASSQAPFIASLCLKSPGRFERILLRAANTHSSAKPCGQQSIHLGGVFELQPGASVFVNVTDPSQVSSpecifications
| CD40LG | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CD40LG. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| P29965 | |
| CD40LG | |
| Recombinant protein corresponding to amino acids of recombinant CD40LG. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| CD40LG | |
| CD154/CD40L/HIGM1/IGM/IMD3/T-BAM/TNFSF5/TRAP/gp39/hCD40L | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 959 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only