Learn More
CD247 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CD247.
Brand: Abnova PAB2494.400uL
Description
The protein encoded by this gene is T-cell receptor zeta, which together with T-cell receptor alpha/beta and gamma/delta heterodimers, and with CD3-gamma, -delta and -epsilon, forms the T-cell receptor-CD3 complex. The zeta chain plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. Low expression of the antigen results in impaired immune response. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
| CD247 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CD247. | |
| In PBS (0.09% sodium azide) | |
| NP_932170;P20963 | |
| CD247 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CD247. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| CD247 | |
| CD3-ZETA/CD3H/CD3Q/CD3Z/T3Z/TCRZ | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 919 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only