Learn More
CD247 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CD247.
Brand: Abnova PAB12667.100ug
Description
The protein encoded by this gene is T-cell receptor zeta, which together with T-cell receptor alpha/beta and gamma/delta heterodimers, and with CD3-gamma, -delta and -epsilon, forms the T-cell receptor-CD3 complex. The zeta chain plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. Low expression of the antigen results in impaired immune response. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
| CD247 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CD247. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| CD3-ZETA/CD3H/CD3Q/CD3Z/T3Z/TCRZ | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes CD247. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. Upon initial thawing apportion into working aliquots and store at -80°C for up to six months. |
| ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) Flow cytometry (10-20 ug per 106 cells) The optimal working dilution should be determined by the end user. | |
| CD247 | |
| CD247 | |
| A synthetic peptide corresponding to extracellular domain of human CD247. | |
| RUO | |
| 919 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only