Learn More
CD163L1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CD163L1.
Brand: Abnova PAB20835.100uL
Description
This gene encodes a member of the scavenger receptor cysteine-rich (SRCR) superfamily. Members of this family are secreted or membrane-anchored proteins mainly found in cells associated with the immune system. The SRCR family is defined by a 100-110 amino acid SRCR domain, which may mediate protein-protein interaction and ligand binding. The encoded protein contains twelve SRCR domains, a transmembrane region and a cytoplasmic domain. Alternatively spliced transcript variants encoding different isoforms have been described but their full-length nature has not been determined. [provided by RefSeq
Sequence: LRVSTRRRGSLEENLFHEMETCLKREDPHGTRTSDDTPNHGCEDASDTSLLGVLPASEATSpecifications
| CD163L1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CD163L1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NR16 | |
| CD163L1 | |
| Recombinant protein corresponding to amino acids of human CD163L1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| CD163L1 | |
| CD163B/M160 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 283316 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only