Learn More
CD14 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CD14.
Brand: Abnova PAB4881.400uL
Description
CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq
Specifications
| CD14 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CD14. | |
| In PBS (0.09% sodium azide) | |
| NP_000582;P08571 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 929 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:50-100) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| CD14 | |
| CD14 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CD14. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only