Learn More
CCNB2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CCNB2.
Brand: Abnova PAB20456.100uL
Description
Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control. [provided by RefSeq
Sequence: TVSSDLENIDTGVNSKVKSHVTIRRTVLEEIGNRVTTRAAQVAKKAQNTKVPVQPTKTTNVNKQLKPTASVKPVQMEKLAPKGPSPTPEDVSMKEENLCQAFSDALLCKIEDIDNEDWENPQLCSpecifications
| CCNB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CCNB2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| O95067 | |
| CCNB2 | |
| Recombinant protein corresponding to amino acids of human CCNB2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| CCNB2 | |
| HsT17299 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9133 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only